D-Luciferin acetate
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
12516
AATB12516EA 307 EUR
AATB12516
D-Luciferin acetate
D-Luciferin acetate
D-Luciferin acetate contains an aceate moiety attached at the 6-O-position, thus preventing it be recognized by the firefly luciferase enzyme.
  • Widely used chemiluminescent substrate
  • Sensitive detection of esterase activity
  • Effective for either lysate samples or in homogenous assays
ADR: 5,II
Order Now
Specificaties
Environmentally Preferable