TF4-VAD-FMK
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
13471
AATB13471EA 152 EUR
AATB13471
TF4-VAD-FMK
TF4-VAD-FMK
TF4-VAD is a red fluorescent cell-permeable polycaspase inhibitor to target caspases 1, 2, 3, 6, 8, 9, or 10.
  • Caspase inhibitor
  • Bright signal
  • Multiple applications
ADR: 5,II