DAX-J2™ Red

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

16301
AATB16301EA 230 EUR
AATB16301
DAX-J2™ Red
DAX-J2™

DAX-J2™ Red is a new nitric oxide (NO) sensor recently developed by AAT Bioquest.


  • Does not require esterase activity for no detection
  • Ph-independent fluorescence
  • More photostable than daf-2 product
  • More sensitive for detecting no than daf-2


Opgelet: For research use only.

ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR