Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 680)

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

16712 16838
AATB16712EA 152 EUR
AATB16712 AATB16838
Anti-IgG Goat Polyclonal Antibody [clone: IgG] (Alexa Fluor® 680)
Antilichamen
AAT Bioquest's iFluor® dyes are optimised for labeling proteins, in particular, antibodies.

  • Bright signal
  • Increased photostability
  • Minimal quenching on proteins

Opgelet: For research use only.

Type: Secondary
Antigen: Immunoglobulin G
Clonality: Polyclonal
Clone: IgG
Conjugation: Alexa Fluor® 680
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR