3'-O-Azidomethyl-dTTP
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17226 17227
AATB17226EA 673 EUR
AATB17226 AATB17227
3'-O-Azidomethyl-dTTP
Reagentia voor nucleïnezuren Nucleotiden
3'-O-Azidomethyl-dTTP is one of the four essential dNTPs (A, C, G, T) that are used for the DNA sequencing by synthesis (SBS).
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Opgelet: For research use only.