dTTP *100 mM PCR Grade*
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17256 17257
AATB17256EA 138 EUR
AATB17256 AATB17257
dTTP *100 mM PCR Grade*
Reagentia voor nucleïnezuren Nucleotiden
2'-Deoxythymidine-5'-Triphosphate (dTTP) solution is used in DNA synthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Opgelet: For research use only.