Acridine orange ≥95%

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Brilliant Acridine Orange E, 3,6-Bis(dimethylamino)acridine, Acridine Orange base, Solvent Orange 15, 3,6-Bis(dimethylamino)acridine N,N,N',N'-Tetramethyl-3,6-acridinediamine, Acridine orange free base, Acridine Orange Stain

17502
AATB17502EA 120 EUR
AATB17502
Acridine orange ≥95%
Acridine orange

Acridine orange is a nucleic acid selective fluorescent cationic dye useful for cell cycle determination.


  • Binding-Determined Bicolor Fluorescence
  • Cell-Permeable


Opgelet: For research use only.

Formule: C₁₇H₁₉N₃
MW: 265.35 g/mol
MDL Nummer: MFCD00005029
CAS nummer: 494-38-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR