Helixyte™ Green *10000X Aqueous PCR Solution*
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17592
AATB17592EA 1480 EUR
AATB17592
Helixyte™ Green *10000X Aqueous PCR Solution*
Testen qPCR doseringen
Helixyte™ Green is manufactured by AAT Bioquest, and it has the same chemical structure of SYBR® Green (SYBR® is the trademark of ThermoFisher).
  • Same Spectral Properties To SYBR® Green
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
Opgelet: For research use only.