Gelite™ 100 bp DNA Ladder
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
17753
AATB17753EA 133 EUR
AATB17753
Gelite™ 100 bp DNA Ladder
Reagentia voor elektroforese Molecuulgewichtmerkers
The Gelite™ 100 bp DNA Ladder is designed for sizing and estimating the mass of double-stranded DNA and PCR products within the 100 to 1500 bp range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
Opgelet: For research use only.