ReadiUse™ TCA Deproteinization Sample Preparation Kit
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
19501
AATB19501EA 230 EUR
AATB19501
ReadiUse™ TCA Deproteinization Sample Preparation Kit
Testen Proteïne-assays
The analysis of amino acids, ions, metabolites and other small molecules is frequently hindered by the interference of lipids, protein and enzymes present in biofluids, cell and tissue lysates.
  • Contains all necessary components
  • Useful for preparing small molecules
  • Can be used in small- and large-scale
Opgelet: For research use only.