Fluo-5F pentapotassium Salt Cell impermeant

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

20562
AATB20562EA 307 EUR
AATB20562
Fluo-5F pentapotassium Salt Cell impermeant
Fluo-5F pentapotassium Salt

Fluo-5F is an analog of Fluo-4 with lower calcium-binding affinity (Kd = ~2.3 µM), making it suitable for detecting intracellular calcium levels in the range of 1 µM to 1 mM that would saturate the response of Fluo-4.


  • Calcium-dependent fluorescence
  • Steady signal
  • Cell impermeant


Opgelet: For research use only.

MW: 931.05 g/mol
ADR: 5 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR