Calbryte™ 590 AM
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
20700 20701 20702
AATB20700EA 307 EUR
AATB20700 AATB20701 AATB20702
Calbryte™ 590 AM
Calbryte™ 590 AM
The intracellular calcium flux assay is a widely used method in monitoring signal transduction pathways and high throughput screening of G protein coupled receptors (GPCRs) and calcium channel targets.
  • Improved background/signal ratio
  • Excellent intracellular retention
  • Long-wavelength emission
  • Cell permeant
Opgelet: For research use only.
ADR: 5,II