FLUO-3 pentasodium salt

Leverancier: AAT BIOQUEST
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

21016
AATB21016EA 230 EUR
AATB21016
FLUO-3 pentasodium salt
FLUO-3 pentasodium salt

Calcium measurement is critical for numerous biological investigations.


  • Well-cited
  • Suitable for all major fluorescence platforms
  • Cell impermeant


Opgelet: For research use only.

Formule: C₃₆H₂₅Cl₂Na₅N₂O₁₃
CAS nummer: 134907-84-9
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR