Psoralen MOP azide
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
39057
AATB39057EA 266 EUR
AATB39057
Psoralen MOP azide
Psoralen MOP azide
Psoralen MOP Azide is an excellent building block for developing site-specific nucleic acid probes via the well-known click chemistry, i.e., Copper-Catalyzed Alkyne–Azide Cycloaddition (CuAAC).
  • Stable for 24 months
Quality tested in-house
Opgelet: For Research Use Only