FMOC-Lys(Cy3)-OH for biomolecular labeling
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
5054
AATB5054EA 1090 EUR
AATB5054
FMOC-Lys(Cy3)-OH for biomolecular labeling
FMOC-Lys(Cy3)-OH
FMOC-Lys(Cy3)-OH is a building block for in-sequence Lys labeling by Cy3.
  • FRET building block
  • Multiple applications
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9016261742b908430ededc1a293383eeecac4a9&vid=0b9ec0a2ef782752d9148a63f4e8510506c18cf7&prid=web&clid=SAPDAM