iFluor® 647, Succinimidyl Ester
Leverancier: AAT BIOQUEST
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1031 71031 71510 71560
AATB1031EA 307 EUR
AATB1031 AATB71031 AATB71510 AATB71560
iFluor® 647, Succinimidyl Ester
iFluor®
AAT Bioquest's iFluor® dyes are optimized for labeling proteins, in particular, antibodies.
  • Bright Fluorescence
  • High Photostability
  • Low Quenching On Proteins
  • pH-Independent Signal
Opgelet: For research use only.