Anti-MAX Rabbit Polyclonal Antibody

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB101271-100
ABCAAB101271-100EA 688 EUR
ABCAAB101271-100
Anti-MAX Rabbit Polyclonal Antibody
Antilichamen
Opgelet:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of

More Product Information
Order Now

Specificaties

antigeen symbool MAX
Type antilichaam Primair,MYC associated factor X
Environmentally Preferable
Type antilichaam Primair,MYC associated factor X
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoNeerslag Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR