Anti-IgG4 Rabbit Monoclonal Antibody [clone: EP4420]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB109493-100
ABCAAB109493-100EA 732 EUR
ABCAAB109493-100
Anti-IgG4 Rabbit Monoclonal Antibody [clone: EP4420]
Antilichamen
Type: Primary
Antigen: IgG4
Clonality: monoclonal
Clone: EP4420
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR