Anti-TIE1 Rabbit Polyclonal Antibody

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB111547-50
ABCAAB111547-50EA 688 EUR
ABCAAB111547-50
Anti-TIE1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: TIE1 (tyrosine kinase with immunoglobulin-like and EGF-like domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR