Creatine Kinase MB (CK-MB) (Rat) ELISA Kit

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB285275-96
ABCAAB285275-96EA 1190 EUR
ABCAAB285275-96
Creatine Kinase MB (CK-MB) (Rat) ELISA Kit
Testen ELISAs

A Sandwich ELISA kit for the quantitative measurement of creatine kinase MB in biological fluids.


  • Detection range: 31,25 to 2000 pg/ml
  • Sensitivity: <18,75 pg/ml
  • Assay precision: Intra-assay: CV <8%; inter-assay: CV <10%
  • No significant cross-reactivity or interference was observed


Easy, convenient and time-saving method to measure the level of creatine kinase MB in serum, plasma, tissue homogenates, culture supernatants and other biological fluids.


Creatine kinase (CK) also known as creatine phosphokinase (CPK) and ATP: creatine N-phosphotransferase is a common cellular enzyme. It catalyses the reversible conversion of creatine and ATP into ADP and phosphocreatine.


CK is widely expressed in various tissues and cell types, with highest activity in striated muscles, heart tissue and brain. CK consists of two subunits: M (muscle) and B (brain), and has three isoenzymes: CK-MM (skeleton muscle), CK-MB (cardiac muscle), and CK-BB (brain).


Increased CK level is associated with many diseases such as myocardial infarction, muscular dystrophy, pulmonary infarction and brain tumors. Accurate measurement of CK is crucial for early diagnosis, prediction and therapeutic strategy. CK-MB ELISA kit is a sandwich ELISA assay for the quantitative measurement of CK-MB in rat serum, plasma and cell culture supernatants. The density of colour is proportional to the amount of CK-MB captured from the samples.


Opgelet:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.


Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR