Anti-NSE Rabbit Polyclonal Antibody

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB834-500
ABCAAB834-500EA 882 EUR
ABCAAB834-500
Anti-NSE Rabbit Polyclonal Antibody
Antilichamen
Opgelet:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of

More Product Information
Order Now

Specificaties

antigeen symbool NSE
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human,Bovine
Host Rabbit
Isotype IgG
ImmunoChemie Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR