Anti-Mumps Nucleoprotein Mouse Monoclonal Antibody [clone: 8H4]

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB9876-1
ABCAAB9876-1EA 625 EUR
ABCAAB9876-1
Anti-Mumps Nucleoprotein Mouse Monoclonal Antibody [clone: 8H4]
Antilichamen
Mouse Monoclonal NCAP antibody. Suitable for ICC/IF and reacts with Mumps orthorubulavirus samples.

Recommended dilutions: ImmunoFluorescence: 1/1 - 1/20.

Type: Primary
Antigen: Mumps
Clonality: Monoclonal
Clone: 8H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR