Anti-CSF1 Rabbit polyclonal antibody unconjugated

Leverancier: Abcam
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB99178-100
ABCAAB99178-100EA 625 EUR
ABCAAB99178-100
Anti-CSF1 Rabbit polyclonal antibody unconjugated
Antilichamen
Opgelet:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of

More Product Information
Order Now

Specificaties

antigeen symbool CSF1
Environmentally Preferable
Type antilichaam Primair,colony stimulating factor 1 (macrophage)
Type antilichaam Primair,colony stimulating factor 1 (macrophage)
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ImmunoChemie Yes
Opslagtemperatuur Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR