Anti-MUC1 Mouse Monoclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AM2009B
ABGEAM2009BEA 354 EUR
ABGEAM2009B
Anti-MUC1 Mouse Monoclonal Antibody
Antilichamen
Type: Primary
Antigen: MUC1 (mucin 1, cell surface associated)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR