Anti-CDKN1A Mouse Monoclonal Antibody [clone: 592CT11.4.4]

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AM2134B
ABGEAM2134BEA 354 EUR
ABGEAM2134B
Anti-CDKN1A Mouse Monoclonal Antibody [clone: 592CT11.4.4]
Antilichamen
Type: Primary
Antigen: CDKN1A
Clonality: Monoclonal
Clone: 592CT11.4.4
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR