Anti-GBA Mouse Monoclonal Antibody [clone: 660CT8.6.6.2]

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AM2180A
ABGEAM2180AEA 354 EUR
ABGEAM2180A
Anti-GBA Mouse Monoclonal Antibody [clone: 660CT8.6.6.2]
Antilichamen
Type: Primary
Antigen: GBA
Clonality: Monoclonal
Clone: 660CT8.6.6.2
Conjugation: Unconjugated
Epitope: internal
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR