Anti-SIGLEC15 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP11503B
ABGEAP11503BEA 354 EUR
ABGEAP11503B
Anti-SIGLEC15 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: SIGLEC15 (sialic acid binding Ig-like lectin 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR