Anti-DLX4 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP14046C
ABGEAP14046CEA 354 EUR
ABGEAP14046C
Anti-DLX4 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: DLX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR