Anti-H Cadherin (CDH13) Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP1434A
ABGEAP1434AEA 354 EUR
ABGEAP1434A
Anti-H Cadherin (CDH13) Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: H Cadherin (CDH13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR