Anti-CARD10 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP14373C
ABGEAP14373CEA 354 EUR
ABGEAP14373C
Anti-CARD10 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CARD10 (caspase recruitment domain family, member 10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR