Anti-AAK1 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP14604C
ABGEAP14604CEA 354 EUR
ABGEAP14604C
Anti-AAK1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: AAK1 (AP2 associated kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR