Anti-CENPU Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP17429B
ABGEAP17429BEA 354 EUR
ABGEAP17429B
Anti-CENPU Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: CENPU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR