Anti-HOXB7 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP18480C
ABGEAP18480CEA 354 EUR
ABGEAP18480C
Anti-HOXB7 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: HOXB7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR