Anti-KIRREL Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP19861B
ABGEAP19861BEA 354 EUR
ABGEAP19861B
Anti-KIRREL Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: KIRREL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR