Anti-KIF9 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP5953B
ABGEAP5953BEA 354 EUR
ABGEAP5953B
Anti-KIF9 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: KIF9 (kinesin family member 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR