Anti-RB1 Rabbit Polyclonal Antibody

Leverancier: Abgent
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AP8575B
ABGEAP8575BEA 354 EUR
ABGEAP8575B
Anti-RB1 Rabbit Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Specificaties

antigeen symbool RB1
Environmentally Preferable
Type antilichaam Primair,Retinoblastoma 1
Antigeen synoniemen RB1|pRb|OSRC|pp110|PP110|P105-RB|RB|Retinoblastoma-associated protein|p105-Rb
Type antilichaam Primair,Retinoblastoma 1
Clonality Polyklonaal
Conjugatie Onconjugeerd
Reactivity Human
Host Rabbit
Gen-ID 3189
Isotype
Western blot Yes
ELISA Yes
ImmunoChemie Yes
Flowcytometrie Yes
Epitoop Center
Immunogeen This HNRNPH3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 90-117 amino acids from the Central region of human HNRNPH3.
Sequentie RLLGQRPGPYDRPC
Purification Protein A & Peptide affinity purification
Concentratie 0,5 mg/ml
Opslagbuffer PBS with 0,09% (W/V) sodium azide, pH 7,4
Opslagtemperatuur Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR