Anti-TNFRSF17 Mouse Monoclonal Antibody [clone: 8H2]

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H00000608-M11
ABNOH00000608-M11EA 397 EUR
ABNOH00000608-M11
Anti-TNFRSF17 Mouse Monoclonal Antibody [clone: 8H2]
Antilichamen
Mouse monoclonal antibody raised against a partial recombinant TNFRSF17.

Type: Primary
Antigen: TNFRSF17
Clonality: Monoclonal
Clone: 8H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a lambda
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR