Anti-RICTOR Mouse Monoclonal Antibody [clone: 1F3]

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

H00253260-M01
ABNOH00253260-M01EA 397 EUR
ABNOH00253260-M01
Anti-RICTOR Mouse Monoclonal Antibody [clone: 1F3]
Antilichamen
Type: Primary
Antigen: RICTOR
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR