Anti-SLC17A6 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S29-29]

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MAB11543
ABNOMAB11543EA 912 EUR
ABNOMAB11543
Anti-SLC17A6 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: S29-29]
Antilichamen
Mouse monoclonal antibody raised against recombinant of Slc17a6.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC17A6
Clonality: Monoclonal
Clone: S29-29
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR