Anti-TNFSF15 Mouse Monoclonal Antibody [clone: VEGI/1283]

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MAB14902
ABNOMAB14902EA 523 EUR
ABNOMAB14902
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: VEGI/1283]
Antilichamen
Mouse monoclonal antibody raised against full length recombinant human TNFSF15.

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: VEGI/1283
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR