Anti-MHVgp1 Mouse Monoclonal Antibody [clone: 2C6.H1]

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

MAB1994
ABNOMAB1994EA 912 EUR
ABNOMAB1994
Anti-MHVgp1 Mouse Monoclonal Antibody [clone: 2C6.H1]
Antilichamen
Mouse monoclonal antibody raised against full length recombinant MHVgp1.

Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:1000

Type: Primary
Antigen: MHVgp1
Clonality: Monoclonal
Clone: 2C6.H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR