Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

P9378.1ML
ABNOP9378.1MLEA 573 EUR
ABNOP9378.1ML
Recombinant mRNA Cap 2'-O-methyltransferase (from E. coli)
Enzymen

Order Now

Specificaties

Enzym naam mRNA Cap 2'-O-methyltransferase
Environmentally Preferable
Enzym type Recombinant
Bron E. coli
Species

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR