Anti-ZFP64 Rabbit Polyclonal Antibody

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PAB22853
ABNOPAB22853EA 523 EUR
ABNOPAB22853
Anti-ZFP64 Rabbit Polyclonal Antibody
Antilichamen
Rabbit polyclonal antibody raised against recombinant ZFP64.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500

Type: Primary
Antigen: ZFP64
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR