Anti-SNX18 Goat Polyclonal Antibody

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PAB6372
ABNOPAB6372EA 523 EUR
ABNOPAB6372
Anti-SNX18 Goat Polyclonal Antibody
Antilichamen
Goat polyclonal antibody raised against synthetic peptide of SNX18.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: SNX18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR