Anti-ADH1A Goat Polyclonal Antibody

Leverancier: Abnova
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

PAB6725
ABNOPAB6725EA 523 EUR
ABNOPAB6725
Anti-ADH1A Goat Polyclonal Antibody
Antilichamen
Goat polyclonal antibody raised against synthetic peptide of ADH1A/ADH1B/ADH1C.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.3-1.5 ?g/ml

Type: Primary
Antigen: ADH1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR