RNase A
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AC118 AC117
AC118EA 184 EUR
AC118 AC117
RNase A
Enzymen
Ribonuclease A (RNase A) is endonuclease-free and quality controlled for use in plasmid purification procedures for digestion of RNA.