Ethyl-4-amino-1-piperidinecarboxylate ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

100240050
ACRO100240050EA 23.5 EUR
ACRO100240050
Ethyl-4-amino-1-piperidinecarboxylate ≥98%
Ethyl-4-amino-1-piperidinecarboxylate

Reactant for synthesis of Milrinone analogs to analyze effects on intracellular calcium increase in cardiac cells


  • Building Block for chemical synthesis

Formule: C₈H₁₆N₂O₂
Kookpunt: 118…120 °C (5mmHg)
Dichtheid: 1,004
Vlampunt: 78 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006484
CAS nummer: 58859-46-4

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4810 to 1.4830 (20 °C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR