3-Chloorpropeen 98% stabilised

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Stabilisatie: Stabilised with propylene oxide

102910010 102910025 102911000
ACRO102910010EA 59.8 EUR
ACRO102910010 ACRO102910025 ACRO102911000
3-Chloorpropeen 98% stabilised
3-Chloorpropeen
Formule: C₃H₅Cl
Kookpunt: 44…46 °C (760 mmHg)
Smeltpunt: –136 °C
Dichtheid: 0,939 g/cm³
Vlampunt: –29 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00000984
CAS nummer: 107-05-1
EINECS: 203-457-6
UN: 1100
ADR: 3,I
Merck Index: 13,00286

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4135 to 1.4175 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.05 to 0.09 % propylene oxide


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR