4-Aminobutyraldehyde diethyl acetal 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: 4,4-Diethoxybutan-1-amine, 4-Aminobutanal diethyl acetal

103260050 103260250 103261000
ACRO103260050EA 34.2 EUR
ACRO103260050 ACRO103260250 ACRO103261000
4-Aminobutyraldehyde diethyl acetal 95%
4-Aminobutyraldehyde diethyl acetal
Formule: C₈H₁₉NO₂
MW: 161.24 g/mol
Kookpunt: 196 °C (760 mmHg)
Dichtheid: 0.9 g/cm³
Vlampunt: 62 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008227
CAS nummer: 6346-09-4
EINECS: 228-753-2
UN: 2735
ADR: 8 III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4270 to 1.4310 (20°C, 589 nm)
GC ≥94 %
Solubility (10% in H2O) Clear colorless solution
Titration with HClO4 99.0 to 101.0 % (On dry substance)
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤200 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR