4-Aminobutyric acid 99+%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: GABA, γ-Aminobutyric acid

103280250 103281000 103285000 103280025
ACRO103280250EA 22.2 EUR
ACRO103280250 ACRO103281000 ACRO103285000 ACRO103280025
4-Aminobutyric acid 99+%
4-Aminobutyric acid
Formule: C₄H₉NO₂
MW: 103.12 g/mol
Kookpunt: 248
Smeltpunt: 197…204 °C (ontbinding)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008226
CAS nummer: 56-12-2
EINECS: 200-258-6
Merck Index: 13,00429

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 197 to 204 °C
Loss on drying ≤0.5 %
Sulfated ash ≤0.1 %
UV transmittance ≥95 % (c=10, 2 N HCl, 430 nm) 1 cm cell
Titration with HClO4 ≥99.0 % (On anhydrous substance)
Chloride (Cl) ≤400 ppm
Sulfate (SO4) ≤500 ppm
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR